Assay Details
Gene Symbol: LOC100060483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607698.3 | XP_005607755.1 | 958 | 96 | ||
XM_005607699.3 | XP_005607756.1 | 958 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
