Assay Details
Gene Symbol: CLDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614708.3 | XP_005614765.1 | 317 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614708.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
