Assay Details
Gene Symbol: MAMSTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729592.2 | XP_014585078.2 | 1669 | 77 | ||
XM_014729598.2 | XP_014585084.1 | 1600 | 77 | |||
XM_014729603.2 | XP_014585089.2 | 1586 | 77 | |||
XM_014729613.2 | XP_014585099.2 | 1522 | 77 | |||
XM_023650054.1 | XP_023505822.1 | 1764 | 77 | |||
XM_023650055.1 | XP_023505823.1 | 1699 | 77 | |||
XM_023650057.1 | XP_023505825.1 | 1695 | 77 | |||
XM_023650058.1 | XP_023505826.1 | 1681 | 77 | |||
XM_023650059.1 | XP_023505827.1 | 1827 | 77 | |||
XM_023650060.1 | XP_023505828.1 | 1475 | 77 | |||
XM_023650061.1 | XP_023505829.1 | 1391 | 77 | |||
XM_023650062.1 | XP_023505830.1 | 1764 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
