Assay Details
Gene Symbol: BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729623.1 | XP_014585109.1 | 1259 | 62 | ||
XM_014729624.1 | XP_014585110.1 | 1256 | 62 | |||
XM_014729625.1 | XP_014585111.1 | 1259 | 62 | |||
XM_014729626.1 | XP_014585112.1 | 1250 | 62 | |||
XM_014729627.2 | XP_014585113.1 | 1175 | 62 | |||
XM_023654238.1 | XP_023510006.1 | 1325 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
