Assay Details
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730027.2 | XP_014585513.1 | 866 | 70 | ||
XM_014730028.2 | XP_014585514.1 | 4934 | 70 | |||
XM_023616027.1 | XP_023471795.1 | 4934 | 70 | |||
XM_023616028.1 | XP_023471796.1 | 4992 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
