Assay Details
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734331.2 | XP_014589817.1 | 135 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014734331.2 transcripts.
Set Membership: |
![]() ![]() |
