Assay Details
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740553.2 | XP_014596039.1 | 314 | 58 | ||
XM_023643055.1 | XP_023498823.1 | 317 | 58 | |||
XM_023643056.1 | XP_023498824.1 | 1155 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
