Assay Details
Gene Symbol: SDR9C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488339.5 | XP_001488389.2 | 5133 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
