Assay Details
Gene Symbol: DDIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488999.4 | XP_001489049.1 | 276 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001488999.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
