Assay Details
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618254.1 | XP_023474022.1 | 243 | 95 | ||
XM_023618255.1 | XP_023474023.1 | 311 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
