Assay Details
Gene Symbol: SUOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491852.6 | XP_001491902.2 | 129 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001491852.6 transcripts.
Set Membership: |
![]() ![]() |
