Assay Details
Gene Symbol: SUOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491852.6 | XP_001491902.2 | 186 | 86 | ||
XM_005611291.3 | XP_005611348.1 | 334 | 86 | |||
XM_005611292.3 | XP_005611349.1 | 268 | 86 | |||
XM_014740898.2 | XP_014596384.1 | 263 | 86 | |||
XM_023643759.1 | XP_023499527.1 | 104 | 86 |
