Assay Details
Gene Symbol: SUOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491852.6 | XP_001491902.2 | 370 | 85 | ||
XM_005611291.3 | XP_005611348.1 | 518 | 85 | |||
XM_005611292.3 | XP_005611349.1 | 452 | 85 | |||
XM_014740898.2 | XP_014596384.1 | 447 | 85 | |||
XM_023643759.1 | XP_023499527.1 | 288 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
