Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491903.5 | XP_001491953.2 | 112 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001491903.5 transcripts.
Set Membership: |
![]() ![]() |
