Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491903.5 | XP_001491953.2 | 714 | 82 | ||
XM_005597141.2 | XP_005597198.1 | 1259 | 82 | |||
XM_023651992.1 | XP_023507760.1 | 858 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
