Assay Details
Gene Symbol: MRPL49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492016.5 | XP_001492066.1 | 412 | 73 | ||
XM_023654422.1 | XP_023510190.1 | 647 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
