Assay Details
Gene Symbol: PTCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492306.6 | XP_001492356.5 | 616 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
