Assay Details
Gene Symbol: IGFLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492598.4 | XP_001492648.1 | 370 | 80 | ||
XM_005596171.3 | XP_005596228.1 | 789 | 80 | |||
XM_023649580.1 | XP_023505348.1 | 789 | 80 | |||
XM_023649581.1 | XP_023505349.1 | 789 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
