Assay Details
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492678.4 | XP_001492728.3 | 367 | 86 | ||
XM_005597198.3 | XP_005597255.1 | 486 | 86 | |||
XM_005597199.3 | XP_005597256.1 | 428 | 86 | |||
XM_014738880.2 | XP_014594366.1 | 374 | 86 | |||
XM_023652082.1 | XP_023507850.1 | 375 | 86 | |||
XM_023652083.1 | XP_023507851.1 | 348 | 86 | |||
XM_023652084.1 | XP_023507852.1 | 374 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
