Assay Details
Gene Symbol: HYAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493703.5 | XP_001493753.3 | 124 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493703.5 transcripts.
Set Membership: |
![]() ![]() |
