Assay Details
Gene Symbol: TIMM17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494341.5 | XP_001494391.1 | 236 | 84 | ||
XM_005614130.3 | XP_005614187.1 | 252 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
