Assay Details
Gene Symbol: SLC5A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496021.5 | XP_001496071.3 | 878 | 60 | ||
XM_005612403.3 | XP_005612460.1 | 878 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
