Assay Details
Gene Symbol: USF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623746.1 | XP_023479514.1 | 400 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023623746.1 transcripts.
Set Membership: |
![]() ![]() |
