Assay Details
Gene Symbol: USF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623746.1 | XP_023479514.1 | 578 | 81 | ||
XM_023623747.1 | XP_023479515.1 | 592 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
