Assay Details
Gene Symbol: LIME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626955.1 | XP_023482723.1 | 435 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
