Assay Details
Gene Symbol: LIME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626955.1 | XP_023482723.1 | 1046 | 75 | ||
XM_023626956.1 | XP_023482724.1 | 1164 | 75 | |||
XM_023626957.1 | XP_023482725.1 | 964 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
