Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629082.1 | XP_023484850.1 | 1464 | 63 | ||
XM_023629091.1 | XP_023484859.1 | 1464 | 63 | |||
XM_023629101.1 | XP_023484869.1 | 1464 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
