Assay Details
Gene Symbol: PDGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631111.1 | XP_023486879.1 | 215 | 65 | ||
XM_023631112.1 | XP_023486880.1 | 215 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
