Assay Details
Gene Symbol: TENT5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633560.1 | XP_023489328.1 | 502 | 107 | ||
XM_023633561.1 | XP_023489329.1 | 595 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023633560.1, XM_023633561.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
