Assay Details
Gene Symbol: PIGV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635387.1 | XP_023491155.1 | 779 | 58 | ||
XM_023635388.1 | XP_023491156.1 | 594 | 58 | |||
XM_023635389.1 | XP_023491157.1 | 305 | 58 | |||
XM_023635390.1 | XP_023491158.1 | 347 | 58 | |||
XM_023635391.1 | XP_023491159.1 | 343 | 58 | |||
XM_023635392.1 | XP_023491160.1 | 1999 | 58 | |||
XM_023635393.1 | XP_023491161.1 | 774 | 58 | |||
XM_023635394.1 | XP_023491162.1 | 896 | 58 | |||
XM_023635395.1 | XP_023491163.1 | 891 | 58 | |||
XM_023635396.1 | XP_023491164.1 | 589 | 58 | |||
XM_023635397.1 | XP_023491165.1 | 711 | 58 | |||
XM_023635398.1 | XP_023491166.1 | 300 | 58 | |||
XM_023635399.1 | XP_023491167.1 | 1995 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
