Assay Details
Gene Symbol: PLA2G2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023635598.1 | XP_023491366.1 | 712 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
