Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636022.1 | XP_023491790.1 | 707 | 57 | ||
XM_023636024.1 | XP_023491792.1 | 572 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
