Assay Details
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642859.1 | XP_023498627.1 | 2177 | 57 | ||
XM_023642860.1 | XP_023498628.1 | 2177 | 57 | |||
XM_023642861.1 | XP_023498629.1 | 2092 | 57 | |||
XM_023642862.1 | XP_023498630.1 | 2176 | 57 | |||
XM_023642863.1 | XP_023498631.1 | 2176 | 57 | |||
XM_023642864.1 | XP_023498632.1 | 2177 | 57 | |||
XM_023642866.1 | XP_023498634.1 | 2177 | 57 | |||
XM_023642867.1 | XP_023498635.1 | 2177 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
