Assay Details
Gene Symbol: GZMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023644216.1 | XP_023499984.1 | 574 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
