Assay Details
Gene Symbol: PRSS57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023644253.1 | XP_023500021.1 | 359 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
