Assay Details
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645219.1 | XP_023500987.1 | 1519 | 59 | ||
XM_023645220.1 | XP_023500988.1 | 1486 | 59 | |||
XM_023645221.1 | XP_023500989.1 | 1483 | 59 | |||
XM_023645222.1 | XP_023500990.1 | 1281 | 59 | |||
XM_023645223.1 | XP_023500991.1 | 1248 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
