Assay Details
Gene Symbol: RNF215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646857.1 | XP_023502625.1 | 693 | 75 | ||
XM_023646859.1 | XP_023502627.1 | 693 | 75 | |||
XM_023646860.1 | XP_023502628.1 | 693 | 75 | |||
XM_023646861.1 | XP_023502629.1 | 693 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
