Assay Details
Gene Symbol: CABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646881.1 | XP_023502649.1 | 268 | 72 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646881.1 | XP_023502649.1 | 268 | 72 |