Assay Details
Gene Symbol: CHST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647597.1 | XP_023503365.1 | 461 | 89 | ||
XM_023647598.1 | XP_023503366.1 | 549 | 89 | |||
XM_023647599.1 | XP_023503367.1 | 2328 | 89 | |||
XM_023647600.1 | XP_023503368.1 | 512 | 89 | |||
XM_023647602.1 | XP_023503370.1 | 419 | 89 | |||
XM_023647603.1 | XP_023503371.1 | 300 | 89 | |||
XM_023647604.1 | XP_023503372.1 | 97 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023647602.1, XM_023647603.1, XM_023647604.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
