Assay Details
Gene Symbol: SNAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655004.1 | XP_023510772.1 | 386 | 98 | ||
XM_023655009.1 | XP_023510777.1 | 383 | 98 | |||
XM_023655015.1 | XP_023510783.1 | 738 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
