Assay Details
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496090.4 | XP_001496140.1 | 421 | 72 | ||
XM_023631046.1 | XP_023486814.1 | 280 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
