Assay Details
Gene Symbol: SLC45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498110.4 | XP_001498160.2 | 917 | 74 | ||
XM_014734912.2 | XP_014590398.1 | 917 | 74 | |||
XM_014734914.2 | XP_014590400.1 | 917 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
