Assay Details
Gene Symbol: NDUFAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498455.5 | XP_001498505.1 | 732 | 72 | ||
XM_023620465.1 | XP_023476233.1 | 727 | 72 | |||
XM_023620466.1 | XP_023476234.1 | 490 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
