Assay Details
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499655.6 | XP_001499705.3 | 311 | 105 | ||
XM_023640307.1 | XP_023496075.1 | 269 | 105 | |||
XM_023640308.1 | XP_023496076.1 | 554 | 105 | |||
XM_023640313.1 | XP_023496081.1 | 235 | 105 | |||
XM_023640316.1 | XP_023496084.1 | 212 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023640307.1, XM_023640308.1, XM_023640313.1, XM_023640316.1 transcripts.
Set Membership: |
![]() ![]() |
