Assay Details
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499655.6 | XP_001499705.3 | 472 | 67 | ||
XM_023640307.1 | XP_023496075.1 | 430 | 67 | |||
XM_023640308.1 | XP_023496076.1 | 715 | 67 | |||
XM_023640313.1 | XP_023496081.1 | 396 | 67 | |||
XM_023640316.1 | XP_023496084.1 | 373 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
