Assay Details
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499655.6 | XP_001499705.3 | 968 | 78 | ||
XM_023640307.1 | XP_023496075.1 | 926 | 78 | |||
XM_023640308.1 | XP_023496076.1 | 1211 | 78 | |||
XM_023640313.1 | XP_023496081.1 | 892 | 78 | |||
XM_023640316.1 | XP_023496084.1 | 869 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
