Assay Details
Gene Symbol: APOA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500785.4 | XP_001500835.1 | 550 | 99 | ||
XM_023644866.1 | XP_023500634.1 | 393 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
