Assay Details
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501392.5 | XP_001501442.3 | 319 | 109 | ||
XM_005602013.2 | XP_005602070.1 | 309 | 109 | |||
XM_005602014.2 | XP_005602071.1 | 295 | 109 | |||
XM_014732895.2 | XP_014588381.1 | 323 | 109 | |||
XM_014732896.2 | XP_014588382.1 | 334 | 109 | |||
XM_014732897.2 | XP_014588383.1 | 314 | 109 | |||
XM_023623802.1 | XP_023479570.1 | 319 | 109 |

More Information
Set Membership: |
![]() ![]() |
