Assay Details
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501392.5 | XP_001501442.3 | 777 | 81 | ||
XM_005602013.2 | XP_005602070.1 | 767 | 81 | |||
XM_005602014.2 | XP_005602071.1 | 753 | 81 | |||
XM_014732895.2 | XP_014588381.1 | 781 | 81 | |||
XM_014732896.2 | XP_014588382.1 | 792 | 81 | |||
XM_014732897.2 | XP_014588383.1 | 772 | 81 | |||
XM_023623802.1 | XP_023479570.1 | 777 | 81 |
