Assay Details
Gene Symbol: KXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503333.5 | XP_001503383.1 | 273 | 70 | ||
XM_023625509.1 | XP_023481277.1 | 365 | 70 | |||
XM_023625510.1 | XP_023481278.1 | 185 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
